Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMDAR1 Splice Variant C2 Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5048P
Description
This Anti-NMDAR1 Splice Variant C2 Antibody is validated for use in immunohistochemistry, western blotting for the detection of NMDAR1 Splice Variant C2.
Specifications
NMDAR1 Splice Variant C2 | |
Polyclonal | |
Affinity purified immunoglobulin. Lyophilized from ammonium bicarbonate (5 mM) so some residual salt may also be present. Reconstitute with 50μL of PBS. Contains no preservatives. | |
Rabbit | |
Affinity Purified | |
Primary | |
Mouse, Rat |
Immunohistochemistry, Western Blot | |
Unconjugated | |
GRIN1; Glutamate; NMDAR1; NMDA1; NR1 | |
Peptide corresponding to the C-terminus of the rat NMDAR1 C2 splice variant. | |
25 μg | |
NM_000832.5; NM_007327.2; NM_021569.2 | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction