Learn More
NMDAR2A&B Rabbit anti-Avian, Guinea Pig, Human, Mouse, Rat, Rabbit, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | NMDAR2A&B |
| Applications | Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified using the immunogen peptide. Lyophilized from PBS containing 1% BSA. Note: lot 0509010940 was shipped in liquid format; Do NOT add additional water to this lot of material. |
| Gene Symbols | GRIN2A; hNR2A; Glutamate; NMDAR2A; NR2A |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide (LNSCSNRRVYKKMPSIESDV) corresponding to the C-terminus of rat NMDAR2A receptor subunit conjugated to BSA. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.