Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR2A Mouse anti-Human, MilliporeSigma™

Catalog No. MAB5216MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5216MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5216MI Supplier MilliporeSigma Supplier No. MAB5216
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-NMDAR2A Antibody detects level of NMDAR2A and has been published and validated for use in immunohistochemistry (paraffin), immunocytochemistry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR2A
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS. Contains no preservative.
Gene Accession No. NM_000833.2
Gene Symbols GRIN2A, hNR2A, Glutamate, NMDAR2A, NR2A
Host Species Mouse
Immunogen Amino acid region 1099-1213 from human NMDAR2A produced as a GST fusion protein.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.