Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR2A Rabbit anti-Human, Mouse, Rat, Fish, Polyclonal, MilliporeSigma™

Catalog No. AB1555MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1555MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1555MI Supplier MilliporeSigma Supplier No. AB1555
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-NMDAR2A Antibody is validated for use in immunoprecipitation, western blotting for the detection of NMDAR2A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR2A
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit antiserum. Liquid, no preservatives.
Gene Symbols GRIN2A; hNR2A; Glutamate; NMDAR2A; NR2A
Host Species Rabbit
Immunogen C-Terminal Fusion Protein of the NMDAR2A (30 kDa), aa 1253-1391.
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000833.2
Target Species Fish, Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.