Learn More
NMDAR2A Rabbit anti-Human, Mouse, Rat, Fish, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | NMDAR2A |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal lyophilized from ammonium bicarbonate (5 mM) so some residual salt may also be present. Reconstitute with 50μL of PBS. Contains no preservative. |
| Gene Symbols | GRIN2A; hNR2A; Glutamate; NMDAR2A; NR2A |
| Host Species | Rabbit |
| Immunogen | C-Terminal Fusion Protein of rat NMDAR2A, aa 1253-1391. |
| Purification Method | Affinity Purified |
| Quantity | 10 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.