Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR2B Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1557PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1557PMI 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1557PMI Supplier MilliporeSigma Supplier No. AB1557P
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-NMDAR2B Antibody is an antibody against NMDAR2B for use in immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR2B
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal lyophilized from ammonium bicarbonate (5 mM). Some residual salt may also be present. Reconstitute with 50μL of PBS. Contains no preservatives.
Gene Symbols GRIN2B; MGC142180; NMDAR2B; NR3; NR2B; MGC142178; hNR3
Host Species Rabbit
Immunogen C-Terminal Fusion Protein of the NMDAR2B (30 kDa).
Purification Method Immunoaffinity purified
Quantity 10 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000834.2
Target Species Human, Mouse, Rat
Content And Storage −20°C for up to 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.