Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMDAR2B Mouse anti-Human, Mouse, Rat, Rabbit, Fish, Clone: 1A8.9B3, MilliporeSigma™

Catalog No. MAB5782MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5782MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5782MI Supplier MilliporeSigma Supplier No. MAB5782
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect NMDAR2B using this Anti-NMDAR2B Antibody, N-terminal validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR2B
Applications Western Blot
Classification Monoclonal
Clone 1A8.9B3
Conjugate Unconjugated
Formulation Liquid ascites fluid
Gene Accession No. NM_000834.2
Gene Symbols GRIN2B, MGC142180, Glutamate, NMDAR2B, NR3, NR2B, MGC142178, hNR3
Host Species Mouse
Immunogen Recombinant protein from rat NMDAR2B.
Purification Method Ascites
Quantity 100 μL
Primary or Secondary Primary
Target Species Fish, Human, Mouse, Rabbit, Rat
Content And Storage −20°C in undiluted aliquots for 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.