Learn More
Description
Specifications
Specifications
| Antigen | NMDAR2C |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized from ammonium bicarbonate (5 mM), some residual salt may be present. Reconstitute with 50μL of PBS. |
| Gene Symbols | GRIN2C; Glutamate; NR2C; NMDAR2C |
| Host Species | Rabbit |
| Immunogen | N-Terminal Fusion Protein of the NMDAR2C |
| Purification Method | Affinity Purified |
| Quantity | 10 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.