Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMT-2 Mouse anti-Human, Unlabeled, Clone: 30, BD

Catalog No. BDB611310 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611310 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611310 Supplier BD Biosciences Supplier No. 611310

Mouse Monoclonal Antibody

Myristoylation is an essential cotranslational modification for many mammalian, viral, and fungal signaling proteins. N-terminal myristoylation is a lipid modification that is catalyzed by N-myristoyltransferase (NMT). NMT transfers myristic acid from myristoyl coenzyme A to the amino group of a protein's N-terminal glycine residue. This modification is important for localization and/or function of many of these proteins. Two human NMTs (NMT-1, NMT-2) have been identified. These proteins share 95% amino acid identity with their mouse homologs. NMT-1 is processed to form four protein isoforms with molecular weights ranging from 49-68kDa. It is not known if these four isoforms are derived from single or multiple gene(s) and their exact functional roles are yet to be determined. In contrast to NMT-1, NMT-2 is a single 65kDa protein. It contains 77% sequence identity with NMT-1, indicating that these proteins comprise two distinct families with overlapping, but distinct, substrate preferences. Additionally, NMT enzymatic activity is increased in colorectal tumors. Thus, NMTs are transferases whose function is essential for biological function of a variety of signaling proteins.

Host Species: Mouse
Clone: 30
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human NMT-2 aa. 10-119
Formula Weight [Chemical]: 65kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen NMT-2
Applications Western Blot
Classification Monoclonal
Clone 30
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human NMT-2 aa. 10-119
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.