Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NO66 Mouse anti-Human, Clone: 8A7-G10, MilliporeSigma™

Catalog No. MABE22MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE22MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE22MI Supplier MilliporeSigma Supplier No. MABE22

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Use Anti-NO66 Antibody, clone 8A7-G10 (Mouse Monoclonal Antibody) validated in western blotting to detect NO66 also known as MYC-associated protein with JmjC domain or Nucleolar protein 66.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NO66
Applications Western Blot
Classification Monoclonal
Clone 8A7-G10
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_076122
Gene Symbols NO66, hsNO66
Host Species Mouse
Immunogen Recombinant protein corresponding to mouse NO66.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.