Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OLFM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528842
Description
Recommended positive controls: HepG2, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Chicken (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The Olfactomedin family comprises a diverse group of secreted glycoproteins, which includes OLFM1 (Noelin-1), OLFM2 (Noelin-2), OLFM3 (Noelin-3), OLFM4 (Noelin-4), tiarin, pancortin, gliomedin and mycocilin. These proteins are implicated in the development of the nervous system. Specifically, OLFM1 and OLFM2 expression is observed in the neural plate and neural crest, as well as in the cranial ganglia in mouse at E8-10, and later in brain tissue and in the zone of polarizing activity in the limb. Overexpression of OLFM1 causes an excess of neural crest emigrations and prolonged neural crest production. OLFM2 participates in the regulation of the development of the anterior nervous system. An Arg144Gln mutation in OLFM2 has been implicated as a possible cause for open-angle glaucoma (OAG).
Specifications
OLFM1 | |
Polyclonal | |
Unconjugated | |
OLFM1 | |
1B426B; AMY; AW742568; D2Sut1e; neuroblastoma protein; neuronal olfactomedin-related ER localized protein; Noe1; Noel; Noel1; Noelin; Noelin 1; Noelin 2; NOELIN1; OlfA; olfactomedin 1; olfactomedin related ER localized protein; olfactomedin-1; OLFM1; Pancortin; pancortin 1; Pancortin 1-4; Pancortin3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10439, 56177 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O88998, Q99784 | |
OLFM1 | |
Synthetic peptide corresponding to a region within amino acids 421 and 485 of Human Noelin. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction