Learn More
nitric oxide synthase 2, inducible, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant NOS2A.
Supplier: Abnova Corporation H00004843A01
Description
Nitric oxide is a reactive free radical which acts as a biologic mediator in several processes, including neurotransmission and antimicrobial and antitumoral activities. This gene encodes a nitric oxide synthase which is expressed in liver and is inducible by a combination of lipopolysaccharide and certain cytokines. Three related pseudogenes are located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq
Sequence: DVRGKQHIQIPKLYTSNVTWDPHHYRLVQDSQPLDLSKALSSMHAKNVFTMRLKSRQNLQSPTSSRATILVELSCEDGQGLNYLPGEHLGVCPGNQPALVQGILERVVDGSpecifications
nitric oxide synthase 2, inducible | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant NOS2A. | |
NOS2 | |
HEP-NOS/INOS/NOS/NOS2A | |
Mouse | |
50 μL | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_000625 | |
NOS2 | |
NOS2A (NP_000616, 685 a.a. ∼ 794 a.a) partial recombinant protein with GST tag. | |
RUO | |
4843 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.