Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOS3, Mouse, Clone: 1D12, Abnova™

Catalog No. 89123249 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-249 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-249 Supplier Abnova Corporation Supplier No. H00004846M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant NOS3.

Nitric oxide is a reactive free radical which acts as a biologic mediator in several processes, including neurotransmission and antimicrobial and antitumoral activities. Nitric oxide is synthesized from L-arginine by nitric oxide synthases. Variations in this gene are associated with susceptibility to coronary spasm. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: QPPEGPKFPRVKNWEVGSITYDTLSAQAQQDGPCTPRRCLGSLVFPRKLQGRPSPGPPAPEQLLSQARDFINQYYSSIKRSGSQAHEQRLQEVEAEVAAT

Specifications

Antigen NOS3
Applications ELISA, In situ PLA, Western Blot
Classification Monoclonal
Clone 1D12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant NOS3.
Formulation PBS with no preservative; pH 7.4
Gene NOS3
Gene Accession No. BC063294
Gene Alias ECNOS/eNOS
Gene Symbols NOS3
Host Species Mouse
Immunogen NOS3 (AAH63294.1, 61 a.a. ∼ 160 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4846
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.