Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOSTRIN, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89203836 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-203-836 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-203-836 Supplier Abnova Corporation Supplier No. H00115677K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human NOSTRIN peptide using ARM Technology.

Nitric oxide (NO) is a potent mediator in biologic processes such as neurotransmission, inflammatory response, and vascular homeostasis. NOSTRIN binds the enzyme responsible for NO production, endothelial NO synthase (ENOS; MIM 163729), and triggers the translocation of ENOS from the plasma membrane to vesicle-like subcellular structures, thereby attenuating ENOS-dependent NO production.[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen NOSTRIN
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description nitric oxide synthase trafficker
Formulation In 1X PBS, pH 7.4
Gene NOSTRIN
Gene Accession No. NOSTRIN
Gene Alias DaIP2, MGC20702
Gene Symbols NOSTRIN
Host Species Rabbit
Immunogen A synthetic peptide of human NOSTRIN
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115677
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.