Learn More
Description
Host Species: Rat
Clone: 22E5.5
Isotype: IgG2a, κ
Cross Reactivity Species: Ms
Immunogen: Mouse Notch1 extracellular domain Recombinant Protein
Flow Cytometry
Specifications
Specifications
| Antigen | Notch 1 |
| Classification | Monoclonal |
| Clone | 22E5.5 |
| Concentration | 0.2mg/mL |
| Conjugate | PE |
| Host Species | Rat |
| Immunogen | Mouse Notch1 extracellular domain Recombinant Protein |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.