Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch 1 Mouse anti-Mouse, Rat, Feline, Clone: mN1A, MilliporeSigma™

Catalog No. MAB5352MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5352MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5352MI Supplier MilliporeSigma Supplier No. MAB5352
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Notch 1 Antibody, clone mN1A is a high quality Mouse Monoclonal Antibody for the detection of Notch 1 and has been validated in immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Notch 1
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone mN1A
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid.
Gene Accession No. NM_017617.3
Gene Symbols NOTCH1, notch1, hN1, TAN1
Host Species Mouse
Immunogen Cdc10-NCR region of mouse Notch1 expressed as a GST fusion protein.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Feline, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.