Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch 1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 071232MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-123-2MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-123-2MI Supplier MilliporeSigma Supplier No. 071232
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Notch 1 Antibody, N-Terminus is a Rabbit Polyclonal Antibody for detection of Notch 1 also known as Neurogenic locus notch homolog protein 1 precursor and has been validated in ELISA, western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Notch 1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal IgG serum in buffer containing 0.02M Potassium Phosphate, 0.15M NaCl, pH 7.2, containing 0.1% sodium azide.
Gene Symbols NOTCH1; notch1; hN1; TAN1
Host Species Rabbit
Immunogen Synthetic peptide from the N-terminal sequence of the cleaved N intracellular domain (NICD) human Notch 1.
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_017617.3
Target Species Human, Mouse
Content And Storage -20°C in undiluted aliquotes for up to 6 months, Avoid Freeze/Thaw Cycles
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.