Learn More
Description
Specifications
Specifications
| Antigen | Notch 2 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Rabbit polyclonal IgG serum in buffer containing 0.02M Potassium phosphate buffer with 0.15M NaCl and 0.1% sodium azide |
| Gene Symbols | NOTCH2; hN2; OTTHUMP00000059536; AGS2 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide from the N-terminal sequence of the cleaved N intracellular domain (NICD) human Notch 2. |
| Quantity | 200 μL |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.