Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch 4 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07189MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-189-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-189-MI Supplier MilliporeSigma Supplier No. 07189
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Notch 4 Antibody is a high quality Rabbit Polyclonal Antibody for the detection of Notch 4 and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Notch 4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, and 0.05% sodium azide.
Gene Symbols NOTCH4; INT3; MGC74442; OTTHUMP00000062678; NOTCH3; hNotch4; notch4; FLJ16302
Host Species Rabbit
Immunogen His-tag fusion protein containing the carboxy-terminus of murine Notch 4
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_004557
Target Species Mouse
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.