Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 071231MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-123-1MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-123-1MI Supplier MilliporeSigma Supplier No. 071231
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-Notch1 Antibody, Intracellular (Rabbit Polyclonal Antibody) validated in ELISA, western blot, immunohistochemistry (paraffin) to detect Notch1 also known as Neurogenic locus notch homolog protein 1 precursor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Notch1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal IgG serum in buffer containing in 0.02M Potassium phosphate, 0.15M NaCl with 0.01% sodium azide.
Gene Symbols NOTCH1; notch1; hN1; TAN1
Host Species Rabbit
Immunogen Synthetic peptide from human Notch 1.
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_017617.3
Target Species Human
Content And Storage -20°C in undiluted aliquotes for up to 6 months, Avoid Freeze/Thaw Cycles
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.