Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch1 Mouse anti-Mouse, PE, Clone: MN1A, BD

Catalog No. BDB552768 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552768 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552768 Supplier BD Biosciences Supplier No. 552768

Mouse Monoclonal Antibody

The Notch family of transmembrane receptors controls cell-fate “decisions” during the development of many organs in a wide variety of animal species. After binding its ligand, a Notch receptor is cleaved in its transmembrane domain, and the resulting intracellular domain dissociates from the membrane and translocates to the nucleus, where it is able to suppress the expression of lineage-specific genes by interacting with transcriptional repressors. The mN1A antibody reacts with the intracellular domain of mouse and human Notch1, but not with mouse Notch2, 3, or 4. In the mouse, Notch1 mRNA is expressed in mouse hematopoietic cells of the fetal liver and adult thymus and bone marrow. Using mAb mN1A, Notch1 is detected in CD4-CD8- (double-negative) and CD4-CD8+ thymocytes. Studies of Notch1-transgenic cells and Notch1-null mice indicate that the receptor is involved in the regulation of lymphopoiesis and myelopoiesis. The mN1A mAb does not cross-react with rat thymocytes.

Host Species: Mouse
Clone: mN1A
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Mouse Notch1 GST fusion protein

Flow Cytometry, Intracellular Staining, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Notch1
Applications Flow Cytometry
Classification Monoclonal
Clone MN1A
Concentration 0.2mg/mL
Conjugate PE
Description Notch1; Notch 1; NOTC1; lin-12; Mis6; Motch A; mT14; N1; NICD; p300; Tan1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse Notch1 GST fusion protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.