Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch3 Mouse anti-Human, PE, Clone: MHN3-21, BD

Catalog No. BDB563593 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563593 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563593 Supplier BD Biosciences Supplier No. 563593
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MHN3-21 monoclonal antibody specifically binds to an extracellular domain of human Notch3. Notch3 is a type 1 transmembrane glycoprotein receptor and member of the Notch family that includes Notch1-Notch4. The Notch3 precursor is cleaved in the Golgi and presents as a cell surface heterodimeric receptor. The Notch3 receptor can bind to several membrane-bound ligands including Jagged1, Jagged2 and Delta1. Upon ligand binding, Notch3 undergoes proteolytic cleavage that results in the release of the Notch intracellular domain, NICD. NICD translocates to the nucleus where it can form a transcriptional activator complex with various transcription factors. These multimeric complexes either positively or negatively regulate the expression of multiple genes including those that orchestrate many facets of embryonic development and the subsequent functioning of organ systems such as the hematopoietic, immune, nervous and cardiovascular systems. Notch3 is reportedly expressed by monocytes but not by CD34+ cells and most types of normal lymphocytes. Notch3 has also been implicated in the development of T cell neoplasia. Notch3 is expressed in vascular smooth muscle cells and plays a key role in neural development. Mutations in Notch3 have been identified as the underlying cause of CADASIL, a cerebral autosomal dominant disease that is the most common form of hereditary stroke disorder.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Notch3
Applications Flow Cytometry
Classification Monoclonal
Clone MHN3-21
Conjugate PE
Description NOTC3; CADASIL; CASIL
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Notch3 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.