Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novel hES cell marker CC9 Mouse anti-Human, Clone: CC9-D6-D5, MilliporeSigma™

Catalog No. MABD70MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABD70MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABD70MI Supplier MilliporeSigma Supplier No. MABD70

Mouse Monoclonal Antibody

Anti-Novel hES cell marker CC9 Antibody, clone CC9-D6-D5 is an antibody against Novel hES cell marker CC9 for use in immunocytochemistry, flow cytometry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Novel hES cell marker CC9
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone CC9-D6-D5
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in PBS with 0.05% sodium azide.
Host Species Mouse
Immunogen Shef-1 human embryonic stem cells
Purification Method Purified by ion exchange chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.