Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novel hES cell marker EF12 Mouse anti-Human, Clone: EF12, MilliporeSigma™

Catalog No. MABD72MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABD72MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABD72MI Supplier MilliporeSigma Supplier No. MABD72

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Detect the Novel hES protein using this Anti-Novel hES cell marker EF12, clone EF12-E6 validated for use in immunocytochemistry and Flow.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Novel hES cell marker EF12
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone EF12
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Host Species Mouse
Immunogen Shef-1 human embryonic stem cells
Purification Method Purified by ion exchange chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.