Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOX4 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABC459MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC459MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC459MI Supplier MilliporeSigma Supplier No. ABC459
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-NOX4 Antibody is a highly specific rabbit polyclonal antibody, that targets NADPH Oxidase (phox) and has been tested in immunohistochemistry and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOX4
Applications Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols NOX4; RENOX
Host Species Rabbit
Immunogen Linear peptide corresponding to the C-terminus of human NOX4.
Purification Method Protein A purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001137308
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.