Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPAT Mouse anti-Human, Unlabeled, Clone: 27, BD

Catalog No. BDB611344 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611344 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611344 Supplier BD Biosciences Supplier No. 611344
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cyclins regulate transitions between cell cycle phases by acting as regulatory subunits of the cyclin-dependent kinases (cdk). The temporal expression of cyclins is tightly regulated and plays a critical role in controlling the enzymatic activity of the cdks. Cyclin-dependent kinase 2 (Cdk2) is a member of a family of cdc2-related cell cycle protein kinases. Cdk2 is expressed earlier in the cell cycle than cdc2 and forms complexes with cyclins A, E, D1, and D3. It is not known if the D cyclins can form active complexes with Cdk2. Cyclin E-Cdk2 kinase is active in the G1 and S phases of the cell cycle and is important, as is Cyclin A-Cdk2, for the progression from G1 to S phase. One substrate for cyclin E-Cdk2 is a Nuclear Protein mapped to the Ataxia Telangiectasia locus, NPAT. This protein associates with cyclin E-Cdk2 and can be phosphorylated by Cdk2. NPAT protein levels peak at the G1/S boundary and overexpression of NPAT accelerates S phase entry, especially after coexpression of cyclin E-Cdk2. Thus, NPAT is a substrate of cyclin E-Cdk2 that may mediate G1 to S phase transition.

Host Species: Mouse
Clone: 27
Isotype: IgG2b
Species Reactivity: Human
Immunogen: Human NPAT aa. 681-803
Formula Weight [Chemical]: 210kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen NPAT
Applications Western Blot
Classification Monoclonal
Clone 27
Concentration 250μg/mL
Conjugate Unconjugated
Description Nuclear Protein Ataxia Telangiectasia
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human NPAT aa. 681-803
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.