Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPFFR2, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002827 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-827 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-827 Supplier Abnova Corporation Supplier No. H00010886B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human NPFFR2 protein.

This gene encodes a member of a subfamily of G-protein-coupled neuropeptide receptors. This protein is activated by the neuropeptides A-18-amide (NPAF) and F-8-amide (NPFF) and may function in pain modulation and regulation of the opioid system. Alternative splicing results in multiple transcript variants. [provided by RefSeq

Sequence: MNEKWDTNSSENWHPIWNVNDTKHHLYSDINITYVNYYLHQPQVAAIFIISYFLIFFLCMMGNTVVCFIVMRNKHMHTVTNLFILNLAISDLLVGIFCMPITLLDNIIAGWPFGNTMCKISGLVQGISVAASVFTLVAIAVDRFQCVVYPFKPKLTIKTAFVIIMIIWVLAITIMSPSAVMLHVQEEKYYRVRLNSQNKTSPVYWCREDWPNQEMRKIYTTVLFANIYLAPLSLIVIMYGRIGISLFRAAVPHTGRKNQEQWHVVSRKKQKIIKMLLIVALLFILSWLPLWTLMMLSDYADLSPNELQIINIYIYPFAHWLAFGNSSVNPIIYGFFNENFRRGFQEAFQLQLCQKRAKPMEAYALKAKSHVLINTSNQLVQESTFQNPHGETLLYRKSAEKPQQELVMEELKETTNSSEI

Specifications

Antigen NPFFR2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human NPFFR2 protein.
Formulation PBS with no preservative; pH 7.4
Gene NPFFR2
Gene Accession No. NM_053036
Gene Alias GPR74/NPFF2/NPGPR
Gene Symbols NPFFR2
Host Species Mouse
Immunogen NPFFR2 (NP_444264.1, 1 a.a. ∼ 420 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10886
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.