Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPM-ALK/ALK Mouse anti-Human, Unlabeled, Clone: ALK1, BD

Catalog No. BDB559254 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559254 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559254 Supplier BD Biosciences Supplier No. 559254

Mouse Monoclonal Antibody

Reacts with NPM-ALK, a chimeric gene product of approximately 80kDa, which is the result of t(2;5)(p23;q35) chromosomal translocation. The gene at 5q35 encodes for the cell cycle-regulated nuclolar protein nucleophosmin (NPM), and the gene at 2p23 encodes a novel receptor tyrosine kinase, anaplastic lymphoma kinase (ALK). The 2;5 translocation results in the expression of NPM-ALK protein, or p80, comprised of the N-terminal portion of NPM (about 40% of the molecule) linked to the entire intracellular portion of ALK. ALK1 antibody recognizes a formalin resistant epitope in both the NPM-ALK chimeric and the 200kDa normal human ALK proteins. Cytoplasmic and nuclear expression of NPM-ALK is seen in t(2;5)+ cell lines SU-DHL-1 and Karpas 299. This antibody is routinely tested by flow cytometric analysis and fluorescence microscopy. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen NPM-ALK/ALK
Applications Flow Cytometry
Classification Monoclonal
Clone ALK1
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen x
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.