Learn More
NPM-ALK Set Mouse anti-Human, PE, Clone: ALK1, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 559257
Description
Reacts with ALK-NPM, a chimeric gene product of approximately 80 kDa, which is the result of t(2;5)(p23;q35) chromosomal translocation. The gene at 5q35 encodes for the cell cycle-regulated nucleolar protein, nucleophism (NPM), while the gene at 2p23 encodes a novel receptor tyrosine kinase, anaplastic lymphoma kinase (ALK). The 2;5 translocation results in the expression of NPM-ALK protein, or p80, comprised of the N-terminal portion of NPM (about 40% of the molecule) linked to the entire intracellular portion of ALK. ALK1 antibody recognizes a formalin resistant epitope in both the NPM-ALK chimeric and the 200 kDa normal human ALK proteins. Cytoplasmic and nuclear expression of NPM-ALK is seen in t(2;5)+ cell lines Su-DHL-1 and Karpas 299.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications
NPM-ALK Set | |
Monoclonal | |
PE | |
Affinity Purified | |
RUO | |
PE Mouse Anti-Human NPM-ALK/ALK (100 tests ), PE Mouse IgG3 Kappa Isotype Control (anti-Fructosan) (100 tests) | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
ALK1 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
IgG3 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.