Learn More
natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptide receptor C), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant NPR3.
Supplier: Abnova Corporation H00004883A01
Description
The family of natriuretic peptides (see MIM 108780) elicit a number of vascular, renal, and endocrine effects that are important in the maintenance of blood pressure and extracellular fluid volume. These effects are mediated by specific binding of the peptides to cell surface receptors in the vasculature, kidney, adrenal, and brain.[supplied by OMIM
Sequence: RQEREALPPQKIEVLVLLPQDDSYLFSLTRVRPAIEYALRSVEGNGTGRRLLPPGTRFQVAYEDSDCGNRALFSLVDRVAAARGAKPDLILGPVCEYAASpecifications
natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptide receptor C) | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant NPR3. | |
NPR3 | |
ANPRC/GUCY2B/NPRC | |
Mouse | |
50 μL | |
Membrane Receptors | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_000908 | |
NPR3 | |
NPR3 (NP_000899, 42 a.a. ∼ 140 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
4883 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.