Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAD(P)H dehydrogenase, quinone 2, Mouse, Clone: 3E8-G2, Abnova™

Catalog No. 89014154 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-154 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-154 Supplier Abnova Corporation Supplier No. H00004835M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant NQO2.

Sequence: MAGKKVLIVYAHQEPKSFNGSLKNVAVDELSRQGCTVTVSDLYAMNFEPRATDKDITGTLSNPEVFNYGVETHEAYKQRSLASDITDEQKKVREADLVIFQFPLYWFSVPAILKGWMDRVLCQGFAFDIPGFYDSGLLQGKLALLSVTTGGTAEMYTKTGVNGDSRYFLWPLQHGTLHFCGFKVLAPQISFAPEIASEEERKGMVAAWSQRLQTIWKEEPIPCTAHWHFGQ

Specifications

Antigen NAD(P)H dehydrogenase, quinone 2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3E8-G2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant NQO2.
Formulation PBS with no preservative; pH 7.4
Gene NQO2
Gene Accession No. BC006096
Gene Alias DHQV/DIA6/NMOR2/QR2
Gene Symbols NQO2
Host Species Mouse
Immunogen NQO2 (AAH06096.1, 1 a.a. ∼ 231 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 4835
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.