Learn More
nuclear receptor subfamily 0, group B, member 1, Mouse, Clone: 1F10, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene encodes a protein that contains a DNA-binding domain. The encoded protein acts as a dominant-negative regulator of transcription which is mediated by the retinoic acid receptor. This protein also functions as an anti-testis gene by acting antagonistically to Sry. Mutations in this gene result in both X-linked congenital adrenal hypoplasia and hypogonadotropic hypogonadism. [provided by RefSeq
Specifications
Specifications
| Antigen | nuclear receptor subfamily 0, group B, member 1 |
| Applications | ELISA, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 1F10 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant NR0B1. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | NR0B1 |
| Gene Accession No. | NM_000475 |
| Gene Alias | AHC/AHCH/AHX/DAX-1/DAX1/DSS/GTD/HHG/NROB1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.