Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
nuclear receptor subfamily 1, group H, member 4, Mouse, Clone: 1B10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant NR1H4.
Supplier: Abnova Corporation H00009971M02
Description
Sequence: TPMFSFYKSIGELKMTQEEYALLTAIVILSPDRQYIKDREAVEKLQEPLLDVLQKLCKIHQPENPQHFACLLGRLTELRTFNHHHAEMLMSWRVNDHKFTPLLCEIWDVQSpecifications
nuclear receptor subfamily 1, group H, member 4 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005123 | |
NR1H4 | |
NR1H4 (NP_005114, 363 a.a. ∼ 472 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Transcription Regulation | |
Primary | |
Human | |
Antibody |
ELISA, Immunofluorescence, Western Blot | |
1B10 | |
Mouse monoclonal antibody raised against a partial recombinant NR1H4. | |
NR1H4 | |
BAR/FXR/HRR-1/HRR1/MGC163445/RIP14 | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
9971 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction