Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NR2A Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07632
Description
Detect NR2A using this Anti-NR2A Antibody validated for use in immunoprecipitation and western blotting.
Specifications
NR2A | |
Polyclonal | |
Purified rabbit polyclonal IgG in buffer containing 70% storage buffer 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide and 30% glycerol. | |
Rabbit | |
Protein A purfied | |
Primary | |
Mouse, Rat | |
Purified |
Immunoprecipitation, Western Blot | |
Unconjugated | |
GRIN2A; hNR2A; Glutamate; NMDAR2A; NR2A | |
6His-tagged fusion protein corresponding to amino acids 1265-1464 of mouse NMDA receptor 2A (NR2A). | |
200 μg | |
NM_008170 | |
−20°C for up to 12 months | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction