Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-NR2A Ab 2, Polyclonal,

Catalog No. AB10531MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10531MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10531MI Supplier MilliporeSigma™ Supplier No. AB10531

Specifications

Antigen NR2A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum containing 0.05% sodium azide.
Gene Symbols GRIN2A; hNR2A; Glutamate; NMDAR2A; NR2A
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to rat NR2A at the C-terminus.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_036705
Target Species Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.