Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR2B Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. 06600MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06600MI 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06600MI Supplier MilliporeSigma Supplier No. 06600
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect NR2B using this Anti-NR2B Antibody validated for use in immunocytochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR2B
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, and 0.05% sodium azide. Frozen solution.
Gene Symbols MGC142178; MGC142180; NMDAR2B; NR3; hNR3; GRIN2B
Host Species Rabbit
Immunogen a.a. 1437-1456 of mature mouse NR2B or 1463-1482 of NR2B precursor; signal is a.a. residues 1-26 [KFNGSSNG HVYEKLSSIESDV]. This sequence is identical to a.a. 1437-1456 of mature rat NR2B and a.a. 1465-1484 of human NR3, containing a N-terminal lysine.
Purification Method Protein A purfied
Quantity 250 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_008171
Target Species Bovine, Canine, Human, Monkey, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.