Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR2B Rabbit anti-Rabbit, Bovine, Canine, Human, Monkey, Rat, FITC, Polyclonal, MilliporeSigma™

Catalog No. FCABS332FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCABS332FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCABS332FMI Supplier MilliporeSigma Supplier No. FCABS332F

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Milli-Mark™ Anti-NR2B-FITC Antibody detects level of NR2B and has been published and validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR2B
Applications Flow Cytometry
Classification Polyclonal
Conjugate FITC
Formulation Purified rabbit polyclonal IgG conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Symbols MGC142178; MGC142180; NMDAR2B; NR3; hNR3; GRIN2B
Host Species Rabbit
Immunogen Linear peptide corresponding to mouse NR2B.
Purification Method Protein A Purified
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) NP_000825
Target Species Bovine, Canine, Human, Monkey, Rabbit, Rat
Content And Storage 2-8°C for 6 months in undiluted aliquotes.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.