Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NR2B Mouse anti-Human, Rat, Clone: BWJHL, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05920
Description
Anti-NR2B Antibody, clone BWJHL detects level of NR2B and has been published and validated for use in western blotting.
Specifications
NR2B | |
Monoclonal | |
Unconjugated | |
GRIN2B, Glutamate, MGC142178, MGC142180, NR2B, NMDAR2B, NR3, hNR3 | |
GST fusion protein corresponding to residues 934-1457 of rat NR2B. Clone BWJHL. | |
100 μg | |
Human, Rat | |
Purified |
Western Blot | |
BWJHL | |
NM_012574 | |
Mouse | |
Protein G purfied | |
Primary | |
−20°C for up to 24 months. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction