Learn More
nuclear receptor subfamily 4, group A, member 3, Mouse, Clone: 1E9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant NR4A3.
Supplier: Abnova Corporation H00008013M02
Description
This gene encodes a member of the steroid-thyroid hormone-retinoid receptor superfamily. The encoded protein may act as a transcriptional activator. The protein can efficiently bind the NGFI-B Response Element (NBRE). Three different versions of extraskeletal myxoid chondrosarcomas (EMCs) are the result of reciprocal translocations between this gene and other genes. The translocation breakpoints are associated with Nuclear Receptor Subfamily 4, Group A, Member 3 (on chromosome 9) and either Ewing Sarcome Breakpoint Region 1 (on chromosome 22), RNA Polymerase II, TATA Box-Binding Protein-Associated Factor, 68kDa (on chromosome 17), or Transcription factor 12 (on chromosome 15). Four transcript variants encoding three distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: LDYSRYCPTDQAAAGTDAEHVQQFYNLLTASIDVSRSWAEKIPGFTDLPKEDQTLLIESAFLELFVLRLSIRSNTAEDKFVFCNGLVLHRLQCLRGFGEWLDSIKDFSSpecifications
nuclear receptor subfamily 4, group A, member 3 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_006981 | |
NR4A3 | |
NR4A3 (NP_008912, 414 a.a. ∼ 521 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Transcription Regulation | |
Primary | |
Human | |
Antibody |
ELISA, Immunofluorescence, Western Blot | |
1E9 | |
Mouse monoclonal antibody raised against a partial recombinant NR4A3. | |
NR4A3 | |
CHN/CSMF/MINOR/NOR1/TEC | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
8013 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.