Learn More
nuclear receptor subfamily 6, group A, member 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human NR6A1 protein.
Supplier: Abnova Corporation H00002649D01P
Description
This gene encodes an orphan nuclear receptor which is a member of the nuclear hormone receptor family. Its expression pattern suggests that it may be involved in neurogenesis and germ cell development. The protein can homodimerize and bind DNA, but in vivo targets have not been identified. The gene expresses at least alternatively spliced transcript variants. [provided by RefSeq
Sequence: MERDEPPPSGGGGGGGSAGFLEPPAALPPPPRNGFCQDELAELDPGTNDRAEQRTCLICGDRATGLHYGIISCEGCKGFFKRSICNKRVYRCSRDKNCVMSRKQRNRCQYCRLLKCLQMGMNRKAIREDGMPGGRNKSIGPVQISEEEIERIMSGQEFEEEANHWSNHGDSDHSSPGNRASESNQPSPGSTLSSRSVELNGFMAFREQYMGMSVPPHYQYIPHLFSYSGHSPLLPQQARSLDPQSYSLIHQLLSAEDLEPLGTPMLIEDGYAVTQAELFALLCRLADELLFRQIAWIKKLPFFCELSIKDYTCLLSSTWQELILLSSLTVYSKQIFGELADVTAKYSPSDEELHRFSDEGMEVIERLIYLYHKFHQLKVSNEEYACMKAINFLNQDIRGLTSASQLEQLNKRYWYICQDFTEYKYTHQPNRFPDLMMCLPEIRYIAGKMVNVPLEQLPLLFKVVLHSCKTSVGKESpecifications
| nuclear receptor subfamily 6, group A, member 1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human NR6A1 protein. | |
| NR6A1 | |
| GCNF/GCNF1/NR61/RTR | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001489.3 | |
| NR6A1 | |
| NR6A1 (NP_001480.3, 1 a.a. ∼ 475 a.a) full-length human protein. | |
| 100 μg | |
| Transcription Regulation | |
| 2649 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.