Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nuclear receptor binding protein 2 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89004829 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-829 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-829 Supplier Abnova Corporation Supplier No. H00340371A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant NRBP2.

Sequence: PRVLAPPPEEVQKAKTPTPEPFDSETRKVIQMQCNLERSEDKARWHLTLLLVLEDRLHRQLTYDLLPTDSAQDLASELVHYGFLHEDDRMKLAAFLESTFLKYRGTQA

Specifications

Antigen nuclear receptor binding protein 2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant NRBP2.
Formulation 50% glycerol
Gene NRBP2
Gene Accession No. NM_178564
Gene Alias DKFZp434P086/MGC138699/TRG16/pp9320
Gene Symbols NRBP2
Host Species Mouse
Immunogen NRBP2 (NP_848659, 151 a.a. ∼ 258 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 340371
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.