Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-NRF-1 Clone: R157.1.3H3,

Catalog No. MABE995MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE995MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE995MI Supplier MilliporeSigma™ Supplier No. MABE995
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects NRF-1 Clone: R157.1.3H3 in Human samples, and it is validated for ChIP, ChIP-seq, Chromatin Immunoprecipitation, Western Blotting

Nuclear respiratory factor 1 (UniProt Q16656; also known as Alpha-pal, Alpha palindromic-binding protein, NRF-1) is encoded by the NRF1 (also known as ALPHA-PAL) gene (Gene ID 4899) in human. NRF-1 is a redox-responsive transcription factor that plays an anti-oxidant, pro-survival role in mitochondrial biogenesis process. NRF-1 is upregulated and activated/phosphorylated by Akt in response to oxidant exposure or increased cellular reactive oxygen species (ROS), resulting in NRF-1 nuclear translocation, where it binds to DNA as a homodimer and mediates the transcription of mitochondrial transcription factor A (mtTFA) that is needed for mitochondrial DNA transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NRF-1
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot, Western Blot
Classification Monoclonal
Clone R157.1.3H3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q16656
Gene Symbols NRF1
Host Species Mouse
Immunogen Recombinant protein corresponding to human NRF-1.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001035199
Target Species Human
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.