Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NSD1 Mouse anti-Rat, Mouse, Clone: 1NW-1A10, MilliporeSigma™

Catalog No. 041565MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-156-5MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-156-5MI Supplier MilliporeSigma Supplier No. 041565
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-NSD1 Antibody, clone 1NW-1A10 is a mouse monoclonal antibody for detection of NSD1, also known as androgen receptor-associated coregulator 267, Lysine N-methyltransferase 3B, has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NSD1
Applications Western Blot
Classification Monoclonal
Clone 1NW-1A10
Conjugate Unconjugated
Formulation Unpurified mouse monoclonal IgG1κ in preservative-free ascites.
Gene Accession No. NP_032765.3
Gene Symbols NSD1, ARA267, H3-K36-HMTase, H4-K20-HMTase, KMT3B, SOTOS, STO
Host Species Mouse
Immunogen Recombinant protein corresponding to mouse NSD1.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.