Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nuclei Mouse anti-Human, Clone: 3E1.3, MilliporeSigma™

Catalog No. MAB4383MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4383MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4383MI Supplier MilliporeSigma Supplier No. MAB4383
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Nuclei Antibody, clone 3E1.3 detects level of Nuclei and has been published and validated for use in flow cytometry, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nuclei
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone 3E1.3
Conjugate Unconjugated
Formulation Liquid in PBS with 0/1% sodium azide
Host Species Mouse
Immunogen Purified Recombinant Human Nuclei
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.