Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nuclei Mouse anti-Human, Cy3, Clone: 235-1, MilliporeSigma™

Catalog No. MAB1281C3MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1281C3MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1281C3MI Supplier MilliporeSigma Supplier No. MAB1281C3
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The anti-Nuclei, clone 235-1, Cy3 conjugate will recognize human cell nuclei only and is useful in xenograph transplantation studies to study stem cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nuclei
Applications Immunocytochemistry
Classification Monoclonal
Clone 235-1
Conjugate Cyanine3
Formulation Purified mouse monoclonal IgG1 conjugated to Cy3 in PBS with 0.1% sodium azide and 15mg/ml BSA.
Host Species Mouse
Immunogen Human nuclei
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from light.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.