Learn More
Invitrogen™ NUP62 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521435
Description
Recommended positive controls: A431, H1299, Neuro 2A, GL261, C8D30, NIH-3T3, BCL-1, Raw264.7, C2C12, PC-12, Rat2. Predicted reactivity: Mouse (95%), Rat (95%), Xenopus laevis (89%), Rhesus Monkey (99%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SDS3 is a subunit of the histone deacetylase.
Specifications
NUP62 | |
Polyclonal | |
Unconjugated | |
Nup62 | |
62 kDa nucleoporin; AA589433; AI426861; AI790512; AU045898; D7Ertd649e; DKFZp547L134; FLJ20822; FLJ43869; IBSN; MGC841; Np62; nuclear pore complex 1; nuclear pore glycoprotein 62; nuclear pore glycoprotein p62; nucleoporin 62; nucleoporin 62kD; nucleoporin 62kDa; nucleoporin Nup62; nucleoporin p62; Nup62; Nupc1; p62; SNDI | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18226, 23636, 65274 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.19 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P17955, P37198, Q63850 | |
Nup62 | |
Recombinant fragment corresponding to a region within amino acids 294 and 522 of nucleoporin p62 (Uniprot ID#P37198). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.