Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleostemin Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA511441
Description
GNL3 may be required to maintain the proliferative capacity of stem cells.
Specifications
Nucleostemin | |
Polyclonal | |
GNL3 | |
Guanine nucleotide-binding protein-like 3; Nucleolar GTP-binding protein 3; Nucleostemin; E2-induced gene 3 protein; Novel nucleolar protein 47; NNP47; E2IG3, NS | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
26354 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q9BVP2 | |
GNL3 | |
KLH conjugated synthetic peptide between 191-220 amino acids from the central region of human Nucleostemin (GNL3) | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction