Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleus and Mitotic Cell Antigen Mouse anti-Human, Clone: A73-B/D12, MilliporeSigma™

Catalog No. MAB3035MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3035MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3035MI Supplier MilliporeSigma Supplier No. MAB3035

Mouse Monoclonal Antibody

Detect Nucleus and Mitotic Cell Antigen with Anti-Nucleus and Mitotic Cell Antigen Antibody, clone A73-B/D12 (Mouse Monoclonal Antibody), that has been shown to work in western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nucleus and Mitotic Cell Antigen
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone A73-B/D12
Conjugate Unconjugated
Formulation Liquid in 0.02 phosphate buffer, 0.25MNaCl pH 7.6 with 0.1% sodium azide.
Host Species Mouse
Immunogen Live Ls 174T cells (colonic carcinoma).
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to 6 months.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.