Learn More
nuclear distribution gene C homolog (A. nidulans), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
NudC was first identified as a regulator of nuclear movement in the asexual reproductive cycle of the filamentous fungus Aspergillus nidulans. Human NUDC is a nuclear movement protein that associates with dynein (see DYNC1H1; MIM 600112) (Aumais et al., 2003 [PubMed 12679384]).[supplied by OMIM
Specifications
Specifications
| Antigen | nuclear distribution gene C homolog (A. nidulans) |
| Applications | Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against a full-length human NUDC protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | NUDC |
| Gene Accession No. | NM_006600 |
| Gene Alias | HNUDC/MNUDC/NPD011 |
| Gene Symbols | NUDC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.