Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUDT1, Rabbit, MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002900 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-900 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-900 Supplier Abnova Corporation Supplier No. H00004521D01
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human NUDT1 protein.

Misincorporation of oxidized nucleoside triphosphates into DNA/RNA during replication and transcription can cause mutations that may result in carcinogenesis or neurodegeneration. The protein encoded by this gene is an enzyme that hydrolyzes oxidized purine nucleoside triphosphates, such as 8-oxo-dGTP, 8-oxo-dATP, 2-hydroxy-dATP, and 2-hydroxy rATP, to monophosphates, thereby preventing misincorporation. The encoded protein is localized mainly in the cytoplasm, with some in the mitochondria, suggesting that it is involved in the sanitization of nucleotide pools both for nuclear and mitochondrial genomes. Several alternatively spliced transcript variants, some of which encode distinct isoforms, have been identified. Additional variants have been observed, but their full-length natures have not been determined. A single-nucleotide polymorphism that results in the production of an additional, longer isoform (p26) has been described. [provided by RefSeq

Sequence: MGASRLYTLVLVLQPQRVLLGMKKRGFGAGRWNGFGGKVQEGETIEDGARRELQEESGLTVDALHKVGQIVFEFVGEPELMDVHVFCTDSIQGTPVESDEMRPCWFQLDQIPFKDMWPDDSYWFPLLLQKKKFHGYFKFQGQDTILDYTLREVDTV

Specifications

Antigen NUDT1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human NUDT1 protein.
Formulation No additive
Gene NUDT1
Gene Accession No. NM_002452
Gene Alias MTH1
Gene Symbols NUDT1
Host Species Rabbit
Immunogen NUDT1 (NP_002443.3, 1 a.a. ∼ 156 a.a) full-length human protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4521
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.